missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ KIT [V654A] Recombinant Human Protein
KIT is a proto-oncogene encoding a transmembrane tyrosine kinase receptor for stem cell factor required for normal hematopoiesis, melangenesis and gametogenesis.
$2650.00 - $11260.00
Specifications
Accession Number | NP_000213, V654A |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris (pH 7.5), 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol |
Molecular Weight (g/mol) | 53.2kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PR7738A
|
Invitrogen™
PR7738A |
100μg |
Each for $2,650.00
|
|
|||||
PV4291
|
Invitrogen™
PV4291 |
1mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Mutations in KIT are associated with gastrointestinal stromal tumors, mast cell disease, acute myelogenous leukemia, and piebaldism. The clinically observed V654A mutant has been demonstrated to be resistant to inhibition with imatinib.
Specifications
NP_000213, V654A | |
50mM Tris (pH 7.5), 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
PDGFR Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
53.2kDa | |
RUO - research use only | |
KIT V654A | |
Human | |
His Tag | |
Frozen |