KIT Recombinant Human Protein DFS Item
KIT Recombinant Human Protein
Manufacturer:  Life Technologies LSPV3589
 View more versions of this product
Catalog No. PV3589
Description
Description
KIT is a proto-oncogene encoding a transmembrane tyrosine kinase receptor for stem cell factor required for normal hematopoiesis, melangenesis and gametogenesis.
Specifications
Specifications
NP_000213
Protein assays and analysis
Frozen
KIT Recombinant Human Protein
1mg
For research use only. Not for use in diagnostic procedures
KIT, cKit, PBT
KIT was pre-diluted in enzyme dilution buffer and assayed in 10mM HEPES (pH 8.0), 10mM MnCl2, 150mM NaCl, 1mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Baculovirus
Amino acids 544-976
49nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 10μg/mL
His Tag
0.42mg/mL
Frozen
53.3kDa
80% by SDS-PAGE
–80°C
1 Each
KIT Recombinant Human Protein
Kinase assay
PDGFR
KIT Recombinant Human Protein
Human
Documents
For Research Use Only. Not for use in diagnostic procedures.