KIT [D816V] Recombinant Human Protein, Invitrogen™ DFS Item
Human KIT, C-terminal fragment, amino acids T544-V976 (as in NCBI/Protein entry NP_000213.1) with a D816V mutation, N-terminal GST-HIS6 fusion protein with a 3C cleavage site, expressed in Sf9 insect cells.
$614.00 - $9,380.00
Specifications
Host Species Human
Protein Tag GST Tag, His Tag
Recombinant Yes
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PV6199 View Documents Life Technologies
LSPV6199
10μg Each for $614.00
PV6200 View Documents Life Technologies
LSPV6200
100μg Each for $2,095.00
PV6201 View Documents Life Technologies
LSPV6201
1mg Each for $9,380.00
Description
Description
  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications
Specifications
Human
GST Tag, His Tag
Yes