missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kinetin, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC226500250
Specifications
Kinetin | |
Glass bottle | |
269.0°C to 271.0°C | |
525-79-1 | |
C10H9N5O | |
15,5359 | |
Solubility in water: soluble. Other solubilities: freely soluble in dilute aquous hcl or naoh,slightly soluble in methanol and ethanol | |
C1=COC(=C1)CNC2=NC=NC3=C2NC=N3 | |
215.21 | |
CHEBI:27407 | |
Fine Crystalline Powder |
Authentic | |
White to Beige | |
25g | |
99% | |
26,IV,3586 | |
6-furfuryladenine, 6-furfurylamino purine, 6-furfurylaminopurine, adenine, n-furfuryl, cytokinin, kinetin, n-furan-2-ylmethyl-9h-purin-6-amine, n-furfuryladenine, n6-furfuryladenine, n6-furfurylamino purine | |
QANMHLXAZMSUEX-UHFFFAOYSA-N | |
N-(furan-2-ylmethyl)-7H-purin-6-amine | |
3830 | |
215.21 | |
99% |
Chemical Identifiers
525-79-1 | |
215.21 | |
6-furfuryladenine, 6-furfurylamino purine, 6-furfurylaminopurine, adenine, n-furfuryl, cytokinin, kinetin, n-furan-2-ylmethyl-9h-purin-6-amine, n-furfuryladenine, n6-furfuryladenine, n6-furfurylamino purine | |
CHEBI:27407 | |
C1=COC(=C1)CNC2=NC=NC3=C2NC=N3 |
C10H9N5O | |
QANMHLXAZMSUEX-UHFFFAOYSA-N | |
3830 | |
N-(furan-2-ylmethyl)-7H-purin-6-amine |
Safety and Handling
Warning
EINECSNumber : 208-382-2