missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kinetin, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 226500010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Kinetin | |
269.0°C to 271.0°C | |
Authentic | |
Glass bottle | |
1 g | |
15,5359 | |
Solubility in water: soluble. Other solubilities: freely soluble in dilute aquous hcl or naoh,slightly soluble in methanol and ethanol | |
C1=COC(=C1)CNC2=NC=NC3=C2NC=N3 | |
215.21 | |
CHEBI:27407 | |
99% |
525-79-1 | |
White to Beige | |
99% | |
C10H9N5O | |
26,IV,3586 | |
kinetin, 6-furfurylaminopurine, 6-furfurylamino purine, 6-furfuryladenine, n6-furfuryladenine, n-furfuryladenine, cytokinin, n-furan-2-ylmethyl-9h-purin-6-amine, n6-furfurylamino purine, adenine, n-furfuryl | |
QANMHLXAZMSUEX-UHFFFAOYSA-N | |
N-(furan-2-ylmethyl)-7H-purin-6-amine | |
3830 | |
215.21 | |
Fine Crystalline Powder |
Chemical Identifiers
525-79-1 | |
215.21 | |
kinetin, 6-furfurylaminopurine, 6-furfurylamino purine, 6-furfuryladenine, n6-furfuryladenine, n-furfuryladenine, cytokinin, n-furan-2-ylmethyl-9h-purin-6-amine, n6-furfurylamino purine, adenine, n-furfuryl | |
CHEBI:27407 | |
C1=COC(=C1)CNC2=NC=NC3=C2NC=N3 |
C10H9N5O | |
QANMHLXAZMSUEX-UHFFFAOYSA-N | |
3830 | |
N-(furan-2-ylmethyl)-7H-purin-6-amine |
Safety and Handling
Warning
EINECSNumber : 208-382-2
RUO – Research Use Only