Please login to your online account to display your discounted pricing
KINEMATICA Polytron™ PT 1600E Benchtop Homogenizers
Fast and successful sample preparation for small-scale applications
$2,359.32 - $2,359.32
Specifications
Includes Control knob and speed scale
Quantity Each
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    
08-451-163
kinematica
11010042
PT1600 E Homogenizer only Each for $2,359.32
Description & Specifications
Specifications
Includes Control knob and speed scale
Quantity Each
The homogenizer processes emulsions, suspensions and foams by reducing the size of solid particles, droplets and gas bubbles to a few microns or less. Small and large amounts of product are dispersed quickly and efficiently.
  • Soft-start with 300w powerful motor
  • Rotor speed range from 5000 up to 30,000rpm
  • Tip speeds up to approx. 14m/s
  • Integrated electronic speed regulation/stabilization with control knob and speed scale
  • Quick coupling for dispersing aggregates and to change heads scatterers without the need for tools
  • Four different, exchangeable dispersing aggregates in EasyClean EC-design
Note: Homogenizer does not come with aggregates
CE marked