Kinematica Polytron™ Model PT 10-35 GT / PT 3100 D Homogenizer Accessory, Generator
Powerful and versatile benchtop homogenizer. KINEMATICA Polytron™ Model PT 10-35 GT-D Homogenizer completely homogenizes samples from 0.1 to 4000mL.
$3,409.73 - $6,754.16
Specifications
For Use With (Equipment) PT 10-35 GT and PT 3100 D
View More Specs
Products 10
Catalog Number Mfr. No. Model Product Type Price Quantity  
Catalog Number Mfr. No. Model Product Type Price Quantity  
08-451-201
View Documents
Kinematica
11030047
PT-DA 12/2EC-B154 Saw Tooth Each for $3,725.05
08-451-199
View Documents
Kinematica
11030010*
PT-DA 05/2EC-B078 Saw Tooth Each for $3,409.73
08-451-200
View Documents
Kinematica
11030026
PT-DA 07/2EC-B101 Saw Tooth Each for $3,877.63
08-451-205
View Documents
Kinematica
11030075
PT-DA 20/2EC-B193 Saw Tooth Each for $4,399.29
08-451-202
View Documents
Kinematica
11030049
PT-DA 12/2MEC-B154 Knife Rotor Each for $3,953.66
08-451-211
View Documents
Kinematica
11030104
PT-DA 30/2MEC-B250 Knife Rotor Each for $5,712.36
08-451-212
View Documents
Kinematica
11030112
PT-DA 30/2ZEC-B250 Z Stator Each for $5,942.25
08-451-213
View Documents
Kinematica
11030110
PT-DA 30/2WEC-B250 W Stator Each for $6,754.16
08-451-214
View Documents
Kinematica
11030114
PT-DA 30/4EC-B250 Fine Emulsion Each for $6,707.05
08-451-207
View Documents
Kinematica
11030080
PT-DA 20/2WEC-B193 W Stator Each for $5,224.27
Description
Description
  • Completely homogenizes samples from 0.1 to 4000mL
  • Powerful 1200W motor provides quiet and reproducible results
  • Homogenize small tissues to fine emulsions and suspensions
  • Get the right results with a variety of generators
Specifications
Specifications
PT 10-35 GT and PT 3100 D