Please login to your online account to display your discounted pricing
KINEMATICA Polytron™ Model PT 10-35 GT / PT 3100 D Homogenizer Accessory, Generator
Powerful and versatile benchtop homogenizer. KINEMATICA Polytron™ Model PT 10-35 GT-D Homogenizer completely homogenizes samples from 0.1 to 4000mL.
Manufacturer: kinematica  11030075
 View more versions of this product
Catalog No. 08-451-205
  • / Each
Description & Specifications
Specifications
Product Type Saw Tooth
For Use With (Equipment) PT 10-35 GT and PT 3100 D
Volume (Metric) Processing 20 to 2000mL
Model PT-DA 20/2EC-B193
  • Completely homogenizes samples from 0.1 to 4000mL
  • Powerful 1200W motor provides quiet and reproducible results
  • Homogenize small tissues to fine emulsions and suspensions
  • Get the right results with a variety of generators