Kinematica™ Polytron™ Homogenizer Stands: Safety Rings
Prevents homogenizer from moving out of position.
Supplier: Kinematica™ 11045035
Description
The safety ring prevents uncontrolled lowering of the drive unit. Can also be used to fixing the drive unit height.Specifications
Safety Positioning Ring | |
Stainless Steel | |
ST-P10/600 stand |