Please login to your online account to display your discounted pricing
KINEMATICA Polytron™ Homogenizer Stands
$1,273.96 - $2,214.86
Specifications
For Use With (Equipment) Homogenizer
View More Specs
Products
Catalog Number Mfr. No. Type Price Quantity    
08-451-73
kinematica
11040010
Handheld Stand Each for $2,214.86
08-451-187
kinematica
11040050
Homogenizer stand; ST-P10/600 Each for $1,273.96
Description & Specifications
Specifications
For Use With (Equipment) Homogenizer
Stands for following Polytron homogenizers:
Handheld
  • PT 2100 E
  • PT 1300 D
Benchtop
  • PT 1600 E
  • PT 2100
  • PT 2500 E
  • PT 4000
  • PT 10-35 GT
  • PT 3100 D
  • PT 6100