Please login to your online account to display your discounted pricing
KINEMATICA Polytron™ EasyClean™ Dispersing Aggregates for PT 2100 Homogenizers
Equipped with saw-teeth, which increases efficiency in preliminary size reduction of droplets/particles and considerably shortens processing time
Manufacturer: kinematica  11030072
 View more versions of this product
Catalog No. 08-451-182
  • / Each
Description & Specifications
Specifications
For Use With (Application) Ideal for dispersing of fibrous, stringy and brittle samples
For Use With (Equipment) Polytron™ PT 2100 homogenizer
Type Generator
Material Stainless Steel
Capacity (Metric) 10 to 2000mL