Please login to your online account to display your discounted pricing
KINEMATICA Polytron™ EasyClean™ Dispersing Aggregates for PT 2100 Homogenizers
Equipped with saw-teeth, which increases efficiency in preliminary size reduction of droplets/particles and considerably shortens processing time
$2,698.11 - $3,732.46
Specifications
Type Generator
For Use With (Equipment) Polytron™ PT 2100 homogenizer
Material Stainless Steel
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) Capacity (Metric) Price Quantity    
08-451-182
kinematica
11030072
Ideal for dispersing of fibrous, stringy and brittle samples 10 to 2000mL Each for $3,732.46
08-451-63
kinematica
11030023*
Ideal for dispersing of cells and tissues in small amount of liquid 0.3 to 10mL Each for $2,698.11
Description & Specifications
Specifications
Type Generator
For Use With (Equipment) Polytron™ PT 2100 homogenizer
Material Stainless Steel
  • Tips of teeth are secured by ring which avoids teeth from bending
  • Made from superior grade stainless-steel
  • Equipped with special slide bearings, which have extraordinary mechanical characteristics and superior chemical resistance
  • Easily dismantled, cleaned, sterilised or autoclaved
Model: PT-DA05/2EC-D066
  • Dispersing of small quantities of plant, animal or human tissues
  • Dissolving solids
Model: PT-DA 07/2EC-D100
  • Dispersing of cell tissues in small amount of liquid
  • Dissolving solids
Model: PT-DA 12/2EC-D148
  • Extraction
  • Producing suspensions and emulsions
Model: PT-DA 20/2EC-D178
  • Producing suspensions and emulsions