Please login to your online account to display your discounted pricing

KINEMATICA Polytron Dispersing Aggregates: for PT-6100D Homogenizers


Manufacturer: kinematica  11030193

 View more versions of this product

Catalog No. 08-451-131

  • / Each
 

Description & Specifications

Specifications

Type Kinematica Polytron Dispersing Aggregates:
For Use With (Equipment) For PT 6100 D homogenizers
Model PT-DA 45/6EC-B250
Sample Volume (English) 200 to 6000mL
Length (Metric) 250mm