missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark Professional™ Kimwipes™ Lens Cleaning Stations

Use anywhere glasses or safety lenses are worn

Manufacturer:  Kimberly-Clark Professional™ KC34623

 View more versions of this product

Catalog No. 29-635-5394



Alert:

Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.


Includes: Wipers and 16 oz. (473mL) of cleaner.

Description

Description

Use anywhere glasses or safety lenses are worn

 

  • Handy one-at-a-time 4.50 × 8.50 in. (11 × 22cm) wipers won't scratch glass or plastic lenses when used wet
  • Silicone-free, antistatic, antifog cleaning solution removes dirt quickly and easily

 

Specifications

Specifications

OSHA
4.4 x 8.4 in. (11.1 x 21.3 cm)
Wipers and 16 oz. (473mL) of Anti-fog, anti-static cleaning solution
Cleaning station
Cleaning Station
Use anywhere glasses or safety lenses are worn
2240/Cs.
SDS
Documents