Kimberly-Clark Professional™ KIMTECH PURE™ Pre-Saturated Wiper Dispenser
One hand, one wiper, one less hassle
Supplier:  Kimberly-Clark Professional™ 76880
Catalog No. 19-166-896
Description
Description
Ideal one-step application of alcohol to critical surfaces. Dispenser offers convenient, one-handed dispensing.
  • Extractor arm automatically lifts wiper
  • Self-closing and self-sealing to prevent wiper dry-out
  • Doesn't generate particles
Specifications
Specifications
Pre-saturated Wiper Dispenser
9 x 7 in. (22.86 x 17.78 cm)
22.86 cm
22.86 cm
For ISO Class 4 or higher cleanroom environments where sterility is required
White
9 in.
7 in.
Polyester
Resealable pouch/case liner