Learn More
Ansell Kimtech™ A6 Liquid Splash Protection Hooded Coveralls
Cleanroom hooded coveralls provide liquid barrier protection with a breathable back panel for use in ISO 7 and 8 environments.
Supplier: Ansell 47685
Description
- Presented unzipped with the inside of the garment first
- Arms telescope folded
- Legs telescope folded with built-in snaps
- Elastic cuffs and thumb loops
- No label on the outer surface
- Hood, front panel, arms and leg made of high performance laminated SMS fabric
- Back panel made of ultralight weight breathable SMS
- 1 large bag of 25 coveralls individually packed per case
Specifications
| Cleanroom Coveralls | |
| SMS | |
| Elastic | |
| Non-sterile | |
| Controlled Environments |
| 2X-Large | |
| Single-use | |
| Hooded | |
| PPE Cat III directive 89/686/EEC, Type 6 limited chemical splash protection; Type 5 particle protection; EN 14126:2003 certified protection against infective agent penetration | |
| Class 10,000-100,000 (ISO 7-8) |