Please login to your online account to display your discounted pricing

Kimble™ White Urea Closures with PTFE-Faced Foam-Backed Rubber Liner


White closures with rubber liner

Manufacturer: kimble™  K73802U8425

 View more versions of this product

Catalog No. 03-340-123A

  • / Case of 432 Each
 

Description & Specifications

Specifications

Closure Liner PTFE/Urea
Color Black
Material Phenolic
No. per Case 144 Cs.
For Use With (Equipment) Bottle
Neck Finish 8-425