Kimble™ Sterile Disposable Polystyrene Milk Bacteriological Pipets
1.1mL polystyrene pipet with plugged mouth end
Manufacturer:  Kimble™ KFS5640011
 View more versions of this product
Catalog No. 12-724-10
Specifications Description & Specifications
Specifications
0.037 oz., 1.1mL
Yes
Plugged mouth end, black scale
0.037 oz.
Polystyrene
Yes
Calibrated To Deliver/Blow Out for measurement of diluted and undiluted milk, as prescribed in Standard Methods for the Analysis of Dairy Products, APHA
0.5, 1, 1.1mL
±0.025mL
1.1mL
Bacteriological Pipet