Please login to your online account to display your discounted pricing

Kimble™ Serum KG-35 Borosilicate Glass Aluminum Seal without Closure


Manufacturer: kimble™  K61000G50

 View more versions of this product

Catalog No. 10-310-240K

  • / Case of 288 Each
 

Description & Specifications

Specifications

Diameter (Metric) 43 mm
Closure Size 20A
Diameter (English) 1.69 in.
Length (Metric) 75mm
Closure Material Aluminum
Material Borosilicate Glass
Capacity (English) 1.69 oz.
Length (English) 2.95 in.
Capacity (Metric) 50mL
Capacity 1.69 oz., 50mL