missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimble™ Serum KG-35 Borosilicate Glass Aluminum Seal without Closure

Neutraglas™

Manufacturer:  Kimble™ K61000G5

 View more versions of this product

Catalog No. 10-310-240J


 

Specifications

Description

Seal, Aluminum; Kimble; Neutraglas; Material: KG-35 Borosilicate Glass; Capacity: 5mL Serum Bottle; O.D.: 23mm; Without closure; Length: 47mm; GPI Neck Finish: 20A

  • No closure
  • GPI Neck Finish: 20A

Description & Specifications

Specifications

0.16 oz., 5mL
0.16 oz.
0.9 in.
Borosilicate Glass
Serum Bottle
23 mm
5mL
Aluminum
1.85 in.
47mm
20A