Please login to your online account to display your discounted pricing

Kimble™ Serum KG-35 Borosilicate Glass Aluminum Seal without Closure


Manufacturer: kimble™  K61000G200

 View more versions of this product

Catalog No. 10-310-240F

  • / Case of 24 Each
 

Description & Specifications

Specifications

Diameter (Metric) 65 mm
Closure Size 20A
Diameter (English) 2.55 in.
Length (Metric) 114mm
Capacity (English) 6.76 oz.
Length (English) 4.48 in.
Material Borosilicate Glass
Capacity (Metric) 200mL
Capacity 6.76 oz., 200mL
Closure Material Aluminum