Please login to your online account to display your discounted pricing

Kimble™ Serum KG-35 Borosilicate Glass Aluminum Seal without Closure


Manufacturer: kimble™  61000G125

 View more versions of this product

Catalog No. 10-310-240C

  • / Case of 144 Each
 

Description & Specifications

Specifications

Capacity (Metric) 125mL
Capacity (English) 4.22 oz.
Closure Size 20A
Diameter (English) 2.12 in.
Diameter (Metric) 54 mm
Length (English) 4.17 in.
Material Borosilicate Glass
Length (Metric) 106mm
Capacity 4.22 oz., 125mL
Closure Material Aluminum