Please login to your online account to display your discounted pricing

Kimble™ Screw Caps for Disposable Glass Tubes


Linerless, one-piece closures with inner sealing ring

Manufacturer: kimble™  73805B13415

 View more versions of this product

Catalog No. 03-340-77C

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Certifications/Compliance ASTM E982
GPI Thread 13-415
For Use With (Equipment) Disposable glass tubes
Color Natural
Material Polypropylene
Thread Size 13-415
Autoclavable Yes
Product Type Screw Cap