Please login to your online account to display your discounted pricing
Kimble™ Pear-Shaped Glass Centrifuge Tubes
$493.90 - $1,361.08
Specifications
Calibration To contain
Capacity (Metric) 100mL
Diameter (Metric) Outside 58mm
Disposable No
Material Borosilicate Glass
View More Specs
Products
Catalog Number Mfr. No. Certifications/Compliance Tolerance Closure Type Graduations Price Quantity    
05-615
kimble™
K45244100
USP, ASTM E 438 0 to 1.5mL in ±0.03mL, >1.5 to 3mL in ±0.2mL, >3 to 5mL in ±0.3mL, >5 to 10mL in ±0.5mL, >10 to 25mL in ±1mL, >25mL in ±2mL Without 0 to 1.5mL in 0.1mL, 1.5 to 5mL in 0.5mL, 5 to 10mL in 1mL, 10 to 25mL in 5mL, 50mL and 100mL Pack of 6 Each for $1,361.08
Case of 2 Pack for $2,722.16
05-624
kimble™
K45220100
USP, ASTM E 438, ASTM D2709 0 to 0.2mL in ±0.01mL, 25mL in ±1mL, >25mL in ±2mL Pennyhead Stopper 0.01mL in 0 to 0.2mL, 25mL, 50mL, and 100mL Pack of 2 Each for $493.90
Case of 3 Pack for $1,481.70
Description & Specifications
Specifications
Calibration To contain
Capacity (Metric) 100mL
Diameter (Metric) Outside 58mm
Disposable No
Material Borosilicate Glass
Max. Force 800xG
Shape Pear
Kimax™ Brand glass. For use in Thermo Scientific™ IEC™ HN-SII Centrifuge with 05-121-1 rotor.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.