DWK Life Sciences Kimble™ Linear HDPE Stoppers
Closed bottom end fits the standard taper neck
Manufacturer:  DWK Life Sciences 28160R27
 View more versions of this product
Catalog No. 14-644-1G
Description
Description
Closed bottom end fits the standard taper neck
  • Heavy-duty Polyethylene stopper
  • Flange on stopper protects glass
Specifications
Specifications
Used as replacement parts for items that are ground to standard taper stopper dimensions
27
Blue,Yellow
Stopper
27
HDPE
Documents