missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimble™ Kontes™ Friedrich Condensers

High-quality, water-cooled condensers to meet your vapor- or air-cooling needs

Manufacturer:  Kimble™ 4562500022

 View more versions of this product

Catalog No. K456250-0022


 

Specifications Description & Specifications

Specifications

0.37cm
45/50
Hose connection sidearm
0.14 in.
Borosilicate Glass
Provide a long vapor path, good heat transfer and anti-flooding characteristics
19cm
Each
13.3 in.
Friedrich Condenser
340mm
7.4 in.