Please login to your online account to display your discounted pricing
Kimble™ Kontes™ Concentrator Tubes for Evaporative Concentrators
Used with Kontes Evaporative Concentrators
$58.49 - $162.41
Specifications
Type Concentrator Tube
View More Specs
Products
Catalog Number Mfr. No. Includes Standard Taper Joint Capacity (Metric) Price Quantity    
K288251-0000
kimble™
2882510000
Without hooks for clamp attachment 14/20 12mL Each for $60.44
K570012-0500
kimble™
5700120500
v 19/22 15mL Each for $58.49
K570050-1025
kimble™
5700501025
Hooks for spring attachment 19/22 10mL Each for $141.85
K570061-0519
kimble™
5700610519
w/o hooks 19/22 10mL Each for $124.10
K570071-1025
kimble™
5700711025
Slow dry tube 19/22 10mL Each for $162.41
K570051-2526
kimble™
5700512526
Slow dry tube 24/25 25mL Each for $157.26
K570051-2525
kimble™
5700512525
Hooks for clamp attachment; 25mL 19/22 10mL Each for $152.05
Description & Specifications
Specifications
Type Concentrator Tube
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.