DWK Life Sciences Kimble™ Kontes™ Center Tear-Off Aluminum Seals, Color Coded
Tamper-evident center tear-off feature protects target area
Supplier: DWK Life Sciences 73821D13
Description
- Allows total removal of seal without decappers
- For use with 13 and 20mm serum finished bottles and vials
- Designed to secure septa and stoppers to bottles and vials
Order Info
Available in blue, red, and green color to help separate and identify samples
Specifications
Green | |
1000 Cs. | |
13 mm |
Aluminum | |
Tear-off |