Please login to your online account to display your discounted pricing
Kimble™ KIMAX™ Reusable Volumetric Class B Pipets
Rugged, color–coded pipets for general laboratory use
$283.80 - $937.20
Specifications
Class B
Material Borosilicate Glass
Product Type Volumetric Pipet
Disposable No
View More Specs
Products
Catalog Number Mfr. No. Graduation Color Length (Metric) Tolerance Capacity (Metric) Price Quantity    
13-649C
kimble™
K370002
Orange 33.5cm ±0.012mL 2mL Case of 12 Each for $316.94
13-649K
kimble™
K3700010
Red 40cm ±0.04mL 10mL Case of 12 Each for $406.71
13-649B
kimble™
K370001
Blue 31cm ±0.012mL 1mL Case of 12 Each for $316.94
13-649D
kimble™
K370003
Black 35cm ±0.02mL 3mL Case of 12 Each for $354.98
13-649F
kimble™
K370005
White 36.5cm ±0.02mL 5mL Case of 12 Each for $283.80
13-649M
kimble™
K3700020
Yellow 50.5cm ±0.06mL 20mL Case of 12 Each for $554.88
13-649N
kimble™
K3700025
Blue 50.5cm ±0.06mL 25mL Case of 12 Each for $587.68
13-649P
kimble™
K3700050
Red 54.5cm ±0.10mL 50mL Case of 12 Each for $663.30
13-649Q
kimble™
K37000100
Yellow 55cm ±0.16mL 100mL Case of 12 Each for $937.20
Description & Specifications
Specifications
Class B
Material Borosilicate Glass
Product Type Volumetric Pipet
Disposable No
  • Epoxy gradations on borosilicate glass for extreme durability
  • Uniform, pulled tip for efficient dispensing
  • Negative graduations to increase functionality
  • Reverse numbering on opposite side of scale for added convenience
  • Cotton plugged and ready to use