DWK Life Sciences Kimble™ KIMAX™ Beaker, Griffin, Low Form
Thick-walled beakers for a standard of durability in the classroom
Manufacturer:  DWK Life Sciences 140001000
 View more versions of this product
Catalog No. S3073010
Description
Description
  • Beaded top, with spout designed to prevent dripping
  • Durable borosilicate glass beakers
  • Graduated scales
  • Large, scratch- and chemical-resistant marking spot
Specifications
Specifications
33.81 oz.
4.21 in.
Excellent Mechanical Strength and Durability, While Providing High Resistance to Chemical Attack and Thermal Shock
100 to 1000 mL
14.5 cm
50 mL
6 Pk.
Griffin
1000 mL
USP, ASTM E438, ASTM E960
10.7 cm
Yes
5.7 in.
Spout
Matte Finish Marking
Dual Scale
Beaker
Borosilicate Glass
Documents