Please login to your online account to display your discounted pricing
Kimble™ KIMAX™ Beaker, Griffin, Low Form
Thick-walled beakers for a standard of durability in the classroom
$80.70 - $189.00
Specifications
Certifications/Compliance USP, ASTM E438, ASTM E960
Material Borosilicate Glass
Product Type Beaker
For Use With (Application) Excellent mechanical strength and durability, while providing high resistance to chemical attack and thermal shock
Graduated Yes
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Price Quantity    
S3073011
kimble™
140001500
1500mL Pack of 4 Each for $110.70
Case of 4 Pack for $442.80
S3073012
kimble™
140002000
2000mL Pack of 4 Each for $189.00
Case of 2 Pack for $378.00
S307302
kimble™
1400030EMW
30mL Pack of 12 Each for $109.35
Case of 4 Pack for $437.40
S3073010
kimble™
140001000
1000mL Pack of 6 Each for $152.55
Case of 4 Pack for $610.20
S307304
kimble™
14000100EM
100mL Pack of 12 Each for $83.05
Case of 4 Pack for $332.10
S307303
kimble™
1400050EMW
50mL Pack of 12 Each for $109.35
Case of 4 Pack for $437.40
S307305
kimble™
14000150EMW
150mL Pack of 12 Each for $106.65
Case of 4 Pack for $426.60
S307306
kimble™
14000250E
250mL Pack of 12 Each for $106.65
Case of 4 Pack for $426.60
S307308
kimble™
14000600E
600mL Pack of 6 Each for $80.70
Case of 6 Pack for $484.00
Description & Specifications
Specifications
Certifications/Compliance USP, ASTM E438, ASTM E960
Material Borosilicate Glass
Product Type Beaker
For Use With (Application) Excellent mechanical strength and durability, while providing high resistance to chemical attack and thermal shock
Graduated Yes
Includes Spout
Label Type Matte finish marking
  • Beaded top, with spout designed to prevent dripping
  • Durable borosilicate glass beakers
  • Graduated scales
  • Large, scratch- and chemical-resistant marking spot