Kimble™ KIMAX™ Addition Funnel with PTFE Stopcock
Supplied with a ground glass stopper and PTFE stopcock
Manufacturer:  Kimble™ K29040F250
 View more versions of this product
Catalog No. 10-310-231D
Specifications Description & Specifications
Specifications
8.45 oz.
PTFE
Addition Funnel
±5mL
250mL, (8.45 oz.)
60mm
USP Type I, ASTM E438 Type I
5mL
2
250mL
2.36 in.
Borosilicate Glass