Please login to your online account to display your discounted pricing

Kimble™ KG-35 Borosilicate Glass Serum Bottle, Aluminum Seal without Closure


Manufacturer: kimble™  K61000G30

 View more versions of this product

Catalog No. 10-310-240H

  • / Case of 288 Each
 

Description & Specifications

Specifications

Closure Material Aluminum
Diameter (English) 1.45 in.
Diameter (Metric) 37mm
Length (English) 2.59 in.
Length (Metric) 66mm
Capacity (Metric) 30mL
Capacity (English) 1.01 oz.
Closure Size 20A
Material Borosilicate Glass
Capacity 1.01 oz., 30mL