Please login to your online account to display your discounted pricing

Kimble™ KG-35 Borosilicate Glass Serum Bottle, Aluminum Seal without Closure


Manufacturer: kimble™  K61000G10

 View more versions of this product

Catalog No. 10-310-240A

  • / Case of 288 Each
 

Description & Specifications

Specifications

Diameter (Metric) 25mm
Closure Size 20A
Length (Metric) 52mm
Diameter (English) 0.98 in.
Closure Material Aluminum
Length (English) 2.04 in.
Capacity (English) 0.33 oz.
Material Borosilicate Glass
Capacity (Metric) 10mL
Capacity 0.33 oz., 10mL