DWK Life Sciences Kimble™ Disposable Polystyrene Serological Pipets in Canister
For mass testing and serological work
Manufacturer:  DWK Life Sciences K5670011100
 View more versions of this product
Catalog No. 13-674-44A
Description
Description
  • Calibrated to deliver to the tip
  • Features two rings near the top
  • Irradiation sterilized
  • Printed with chemically resistant organic black ink
Specifications
Specifications
Yes
0.03 oz., 1mL
For tissue culture work, mass testing and serological work
0.01mL
Canister
±0.02mL
1mL
Sterile
0.03 oz.
Yes
Yes
0.3mL
500/Cs.
Serological Pipet
Plastic
Documents