DWK Life Sciences Kimble™ Disposable Plastic Standard-Tip Serological Pipets in Multipacks
Irradiation sterilized and suitable for tissue culture work
Manufacturer:  DWK Life Sciences K568001110
 View more versions of this product
Catalog No. 13-674-40
Description
Description
  • Have uniform pulled or molded tips and beveled or molded top ends with cotton plugging
  • Two rings near top denote that small amount remaining in tip after delivery must be blown out and added to main volume delivered
  • 5mL size now has two-piece design
  • Color-coded packaging makes choosing correct pipet size quick and accurate
  • Calibrated “To Deliver” to tip
Order Info
Multipacked 10mL pipets available with either regular tip (1.5mm dia.) or wide tip (3.5mm dia.). All sizes available either in paper/plastic individual wraps overpacked in dispensing shelf packs, or in recloseable multipacks overpacked in shipping containers.
Compliance
Designed from ASTM™ E 934.
Specifications
Specifications
Yes
0.03 oz.
ASTM E934
Yes
Yes
0.3mL
1000/Cs.
Serological Pipet
Plastic
Yes
0.03 oz., 1mL
Yellow
For tissue culture work
0.1mL
Bag of 25
Standard, with cotton plugs
1mL
Sterile
Documents