Please login to your online account to display your discounted pricing

Kimble™ Disposable Micro Capillary Pipets


Color-coded for selection of correct size

Manufacturer: kimble™  K71900100

 View more versions of this product

Catalog No. K71900-100

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Certifications/Compliance ASTM , E672, USP type III
Color Blue
Disposable Yes
Length (English) 5 in.
Length (Metric) 127mm
Tolerance (Metric) ±0.5mL
Capacity (Metric) 100μL
Material Soda Lime Glass
Product Type Micro Capillary Pipet