DWK Life Sciences Kimble™ Disposable Individually-Wrapped Plastic Serological Pipets
Irradiation sterilized and suitable for tissue culture work
Manufacturer:  DWK Life Sciences K5690011100
 View more versions of this product
Catalog No. 13-674-41A
Description
Description
  • Have uniform pulled or molded tips and beveled or molded top ends with cotton plugging
  • Two rings near top denote that small amount remaining in tip after delivery must be blown out and added to main volume delivered
  • 5mL size now has two-piece design
  • Color-coded packaging makes choosing correct pipet size quick and accurate
  • Calibrated “To Deliver” to tip
Order Info
Multipacked 10mL pipets available with either regular tip (1.5mm dia.) or wide tip (3.5mm dia.). All sizes available either in paper/plastic individual wraps overpacked in dispensing shelf packs, or in recloseable multipacks overpacked in shipping containers.
Compliance
Designed from ASTM™ E 934.
Specifications
Specifications
Yes
ASTM E934
Yes
0.01mL
Individually wrapped
±0.02mL
1mL
Sterile
Yes
Yellow
Yes
0.3mL
200/Pk.; 1000/Cs.
Serological Pipet
Plastic
Documents
Accessories