Kimble™ Disposable Borosilicate Glass Serological Pipets
Sterile, plugged short-length pipets perfect for tissue culture work or procedures performed under fume hoods or in confined areas
Manufacturer:  Kimble™ K7211025210
 View more versions of this product
Catalog No. 03-340-126D
Specifications Description & Specifications
Specifications
Green
Yes
Serological Pipet
USP type I, ASTM E438
25mL
±1mL
Yes
0.2mL
Borosilicate Glass
5mL
Accessories