Kimble™ Disposable Borosilicate Glass Serological Pipets
Sterile, plugged short-length pipets perfect for tissue culture work or procedures performed under fume hoods or in confined areas
Manufacturer:  Kimble™ K721105110
 View more versions of this product
Catalog No. 03-340-126B
Specifications Description & Specifications
Specifications
2mL
Yes
USP type I, ASTM E438
Yes
±0.2mL
5mL
Borosilicate Glass
Serological Pipet
0.1mL
Blue
Accessories