Kimble™ KIMAX™ Conical-Bottom Glass Centrifuge Tubes: 50mL Capacity
$298.57 - $1,129.20
Specifications
Certifications/Compliance USP, ASTM E 438
Max. Force 2050xG
Shape Conical
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Closure Type Disposable Graduations Material Price Quantity  
Catalog Number Mfr. No. Closure Type Disposable Graduations Material Price Quantity  
05-538-29B Kimble™
K4517450
No Heavy-duty Kimax Brand Case of 6 Each for $550.03
05-538-32C Kimble™
K4516650
No Kimax Brand Case of 12 Each for $1,129.20
05-538-33C Kimble™
K4516750
No Kimax Brand Case of 12 Each for $775.91
05-538-40B Kimble™
K4517650
No Heavy-duty Kimax Brand Case of 6 Each for $812.91
05-538-41A Kimble™
K7378550
Yes Borosilicate Glass Case of 72 Each for $298.57
05-538-41E Kimble™
K7379050
Yes Borosilicate Glass Case of 72 Each for $318.05
05-569-6C Kimble™
K4516550
No Kimax Brand Case of 12 Each for $795.34
Specifications
Description
  • With conical shaped bottom
  • Tubes are made from borosilicate glass in compliance with ASTM Specification E438, Type I, Class A or B glass requirements
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
USP, ASTM E 438
2050xG
Conical