missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ KIMAX™ Conical-Bottom Glass Centrifuge Tubes: 15mL Capacity

Withstand centrifugation up to 2980 RCF

Manufacturer:  DWK Life Sciences 4516615

 View more versions of this product

Catalog No. 05-538-32B



Description

Description

Withstand centrifugation up to 2980 RCF

  • With conical shaped bottom
  • Tubes are made from borosilicate glass in compliance with ASTM Specification E438, Type I, Class A or B glass requirements

Specifications

Specifications

With screw cap
0.1mL
2,980 x g
USP, ASTM E 438
No
Marking Spot
Conical
0 to 0.2mL in ±0.05mL, >0.2 to 1mL in ±0.1mL, >1 to 3mL in ±0.15mL, >3mL in ±0.2mL
17mm
130mm
To contain
15mL
Borosilicate Glass
PTFE
15-415
Centrifuge Tube
Documents