Please login to your online account to display your discounted pricing
Kimble™ KIMAX™ Conical-Bottom Glass Centrifuge Tubes: 100mL Capacity
$721.94 - $1,560.32
Specifications
Capacity (Metric) 100mL
Diameter (Metric) Outside 45mm
Material Borosilicate Glass
Max. Force 800xG
For Use With (Application) Used in the determination of residues in liquified petroleum (LP) gases
View More Specs
Products
Catalog Number Mfr. No. Closure Type Diameter (Metric) Outside Graduations Length (Metric) Price Quantity    
05-605
kimble™
K45240100
Without 37mm 0 to 0.5mL in 0.05mL, 0.5 to 2mL in 0.1mL, 2 to 3mL in 0.2mL, 3 to 5mL in 0.5mL, 5 to 10mL in 1mL, 10 to 25mL in 5mL, 25 to 100mL in 25mL 203mm Pack of 6 Each for $721.94
Case of 2 Pack for $1,443.88
05-621
kimble™
K45243100
Without 45mm 0 to 0.5mL in 0.05mL, 0.5 to 2mL in 0.1mL, 2 to 3mL in 0.2mL, 3 to 5mL in 0.5mL, 5 to 10mL in 1mL, 10 to 25mL in 5mL, and at 50mL, 100mL 165mm Pack of 6 Each for $932.68
Case of 2 Pack for $1,865.36
05-606
kimble™
K45241100
Without 37mm 0 to 1mL in 0.05mL, 1 to 3mL in 0.1mL, 3 to 6mL in 0.2mL, 6 to 10mL in 0.5mL, 10 to 100mL in 1mL 203mm Pack of 6 Each for $875.91
Case of 2 Pack for $1,751.82
05-605-001
kimble™
K45239100
Snap 38mm 0 to 0.5mL in 0.05mL, 0.5 to 2mL in 0.1mL, 2 to 3mL in 0.2mL, 3 to 5mL in 0.5mL, 5 to 10mL in 1mL, 10 to 25mL in 5mL, and at 50mL, 75mL, 100mL 200mm Case of 12 Each for $1,560.32
Description & Specifications
Specifications
Capacity (Metric) 100mL
Diameter (Metric) Outside 45mm
Material Borosilicate Glass
Max. Force 800xG
For Use With (Application) Used in the determination of residues in liquified petroleum (LP) gases
Shape Conical
Tolerance 0 to 0.1mL in ±0.02mL, >0.1 to 0.3mL in ±0.03mL, >0.3 to 0.5mL in ±0.05mL, >0.5 to 1mL in ±0.075mL, >1 to 1.5mL in ±0.1mL, >1.5 to 2mL in <special id=
  • With conical shaped bottom
  • Tubes are made from borosilicate glass in compliance with ASTM Specification E438, Type I, Class A or B glass requirements
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.