Kimble™ Clear Glass Graduated AC Medium Rounds without Caps
Wide mouth design for efficient addition or removal of contents
$221.97 - $642.62
Specifications
Closure Type Screw
Color Clear
Material Glass
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity Closure Size Diameter (English) Height (English) Price Quantity  
Catalog Number Mfr. No. Capacity Closure Size Diameter (English) Height (English) Price Quantity  
13-757-028 Dwk Life Sciences Llc
K5810133B
1 oz., 30mL 33-400 1.33 in. 2.67 in. Case of 432 Each for $642.62
13-757-031 Kimble™
K5810858B
8 oz., 250mL 58-400 2.44 in. 5 in. Case of 96 Each for $232.21
13-757-032 Kimble™
K5811670B
16 oz., 500mL 70-400 2.99 in. 5.7 in. Case of 48 Each for $221.97
13-757-029 Kimble™
K5810238B
2 oz., 60mL 38-400 1.65 in. 3.26 in. Case of 288 Each for $364.00
13-757-030 Kimble™
K5810448B
4 oz., 125mL 48-400 2 in. 4.01 in. Case of 144 Each for $285.54
Specifications
Description
  • AC Medium Round bottles are ideal for storage for solids, liquids, powders and specimens
  • Clear glass allows for easy viewing of contents
  • Choose from a variety of cap / liner combinations in Bulk Packs
  • Manufactured from soda-lime glass conforming to USP Type III and ASTM E438, Type II requirements
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Screw
Clear
Glass