Kimble™ Clear Glass AC Medium Rounds with PTFE-Faced LDPE Foam-lined Caps
Wide mouth design for efficient addition or removal of contents
$394.03 - $1,045.64
Specifications
Closure Liner PTFE/LDPE
Closure Material Phenolic
Closure Type Screw
Closure Color Black
Color Clear
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity Closure Size Diameter (English) Height (English) Price Quantity  
Catalog Number Mfr. No. Capacity Closure Size Diameter (English) Height (English) Price Quantity  
13-756-431 Kimble™
K5710448C26
4 oz., 125mL 48-400 2 in. 4.01 in. Case of 144 Each for $681.95
13-756-430 Kimble™
K5710238C26
2 oz., 60mL 38-400 1.65 in. 3.26 in. Case of 288 Each for $1,045.64
13-756-433 Kimble™
K5711670C26
16 oz., 500mL 70-400 2.99 in. 5.7 in. Case of 48 Each for $394.03
13-756-432 Kimble™
K5710858C26
8 oz., 250mL 58-400 2.44 in. 5 in. Case of 96 Each for $649.62
Specifications
Description
  • AC Medium Round bottles are ideal for storage for solids, liquids, powders and specimens
  • Clear glass allows for easy viewing of contents
  • Choose from a variety of cap / liner combinations in Bulk Packs
  • Manufactured from soda-lime glass conforming to USP Type III and ASTM E438, Type II requirements
Description & Specifications
Specifications
PTFE/LDPE
Screw
Clear
Phenolic
Black
Glass