DWK Life Sciences Kimble™ ValueWare Buret with Threaded Closure Fit
Designed with threaded closure, which eliminates accidental dislodging of the stopcock assembly during use.
$139.18 - $160.23
Specifications
Certifications/Compliance Class B
Type Buret
Graduations 0.1 mL
Includes Straight bore stopcock, dust cap, threaded closure fit
Stopcock Material Glass
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Length (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Length (Metric) Price Quantity  
S01518 View Documents DWK Life Sciences
57121H25
25 mL 42 cm Each for $139.18
S01519 View Documents DWK Life Sciences
57121H50
50 mL 72.5 cm Each for $160.23
Description
Description
  • With straight bore
  • Modular design
  • Dust cap included
  • Assembly should be wetted before installation
  • Ground tip fits into lower end of PTFE stopcock
Specifications
Specifications
Class B
0.1 mL
Glass
Buret
Straight bore stopcock, dust cap, threaded closure fit