Please login to your online account to display your discounted pricing

Kimble™ Straight-Sided Serum Stoppers


For research and pharmaceutical packaging applications

Manufacturer: kimble™  K7381113

 View more versions of this product

Catalog No. 03-340-301

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Certifications/Compliance USP, EP, JP
For Use With (Application) For research and pharmaceutical packaging applications
For Use With (Equipment) 13mm GPI aluminum seal finish
Hardness 50 Durometer
Latex Free Yes
Color Gray
Material Chlorobutyl
Product Type Straight-Sided Serum Stopper