Please login to your online account to display your discounted pricing
Open-Top PolypropyleneScrew Thread Closures with Red PTFE-Faced Silicone Septa
Autoclavable and used for multiple injections
Manufacturer: kimble chase life science  K73812BL9425
 View more versions of this product
Catalog No. 03-341-610
  • / Case of 1000 Each
Description & Specifications
Specifications
Type Open Top Screw Cap
Material Polypropylene
GPI Thread 9-425
Autoclavable Yes
Color Blue
Material Closure Polypropylene
Pre-slit No
Thread Size 9-425