Please login to your online account to display your discounted pricing
Kimble™ Open-Top PolypropyleneScrew Thread Closures with Bonded White PTFE Liners
Intended for single-use injection
Manufacturer: kimble™  K73814WH8425
 View more versions of this product
Catalog No. 03-341-616
  • / Case of 1000 Each
Description & Specifications
Specifications
Color White
Material Polypropylene
GPI Thread 8-425
Type Open Top Screw Cap
Material Closure Polypropylene
Material (Liner) PTFE
Autoclavable Yes
Thread Size 8-425