missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Kimble™ Open-Top Aluminum Seals, Color Coded
Designed to secure septa and stoppers to bottles and vials
$226.18 - $353.60
Catalog Number | Mfr. No. | Color | Thread Size | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Thread Size | Price | Quantity | ||||
03-340-305
|
DWK Life Sciences
73822A13 |
Natural | 13 mm |
Case of 1000 Each for $226.18
|
|
03-340-309
|
DWK Life Sciences
73822B20 |
Blue | 20 mm |
Case of 1000 Each for $243.04
|
|
03-340-307
|
DWK Life Sciences
73822C13 |
Red | 13 mm |
Case of 1000 Each for $261.16
|
|
03-340-308
|
DWK Life Sciences
73822D13 |
Green | 13 mm |
Case of 1000 Each for $245.22
|
|
03-340-310
|
DWK Life Sciences
73822C20 |
Red | 20 mm |
Case of 1000 Each for $353.60
|
|
03-340-311
|
DWK Life Sciences
73822D20 |
Green | 20 mm |
Case of 1000 Each for $332.03
|
|
Description
- For use with 13 and 20mm serum finished bottles and vials
- Available in blue, red, green and natural aluminum color to help separate and identify samples
Specifications
Open Top | |
Aluminum | |
1000 Case |